siRNA Details
| Gene Symbol: OR52K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005171.2 | NP_001005171.2 | 1 | 38 | ||
| GenBank mRNA | BC137056.1 | - | 1 | 38 | 9021053 | |
| BC137059.1 | - | 1 | 38 | 9021056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR52K1| Gene Symbol: OR52K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005171.2 | NP_001005171.2 | 1 | 38 | ||
| GenBank mRNA | BC137056.1 | - | 1 | 38 | 9021053 | |
| BC137059.1 | - | 1 | 38 | 9021056 | ||