siRNA Details
| Gene Symbol: OR56A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003443.2 | NP_001003443.2 | 1 | 601 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR56A3| Gene Symbol: OR56A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003443.2 | NP_001003443.2 | 1 | 601 | ||