siRNA Details
| Gene Symbol: OR5T1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004745.1 | NP_001004745.1 | 1 | 17 | ||
| GenBank mRNA | BC136992.1 | - | 1 | 103 | 9020984 | |
| BC136993.1 | - | 1 | 103 | 9020985 | ||
More Information
Set Membership: |
|

OR5T1
Genomic Map