siRNA Details
Gene Symbol: OR6X1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005188.1 | NP_001005188.1 | 1 | 30 | ||
GenBank mRNA | BC136855.1 | - | 1 | 57 | 9020846 | |
BC136856.1 | - | 1 | 57 | 9020847 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OR6X1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005188.1 | NP_001005188.1 | 1 | 30 | ||
GenBank mRNA | BC136855.1 | - | 1 | 57 | 9020846 | |
BC136856.1 | - | 1 | 57 | 9020847 |