siRNA Details
| Gene Symbol: MYO18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_078471.3 | NP_510880.2 | 39 | 5979 | ||
| NM_203318.1 | NP_976063.1 | 39 | 5979 | |||
| GenBank mRNA | AB177858.1 | - | 38 | 5798 | ||
| AB177860.1 | - | 39 | 4931 | |||
| AF252258.1 | - | 2 | 277 | |||
| AK122870.1 | - | 6 | 1115 | |||
| AK294180.1 | - | 6 | 854 | |||
| AK302505.1 | - | 38 | 4705 | |||
| AK302725.1 | - | 23 | 2693 | |||
| AY703984.1 | - | 39 | 4931 | |||
| BC039612.1 | - | 38 | 5972 | 6045482 | ||
| CR933614.1 | - | 39 | 5954 | |||
| D86970.2 | - | 39 | 6002 | |||

MYO18A
Genomic Map