siRNA Details
| Gene Symbol: METTL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304467.1 | NP_001291396.1 | 3 | 350 | ||
| NM_001304468.1 | NP_001291397.1 | 3 | 350 | |||
| NM_212554.3 | NP_997719.2 | 3 | 350 | |||
| GenBank mRNA | AF318345.1 | - | 3 | 278 | ||
| AL832292.1 | - | 3 | 247 | |||
| BC026167.1 | - | 3 | 284 | 4823003 | ||
| BI551093.1 | - | 3 | 244 | |||
| HY048148.1 | - | 3 | 351 | |||

METTL10
Genomic Map