siRNA Details
| Gene Symbol: DNAH10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207437.3 | NP_997320.2 | 16 | 2417 | ||
| XM_005253555.3 | XP_005253612.1 | 17 | 3080 | |||
| XM_011538014.2 | XP_011536316.1 | 17 | 3080 | |||
| XM_011538015.2 | XP_011536317.1 | 17 | 3080 | |||
| XM_011538016.2 | XP_011536318.1 | 17 | 3080 | |||
| XM_011538017.2 | XP_011536319.1 | 17 | 3080 | |||
| XM_011538020.2 | XP_011536322.1 | 17 | 3088 | |||
| XM_017018960.1 | XP_016874449.1 | 17 | 3080 | |||
| XM_017018961.1 | XP_016874450.1 | 17 | 3080 | |||
| GenBank mRNA | AK125475.1 | - | 11 | 1931 | ||
| BC144575.1 | - | 12 | 2078 | |||
| BC150622.1 | - | 12 | 2078 | |||

DNAH10
Genomic Map