siRNA Details
| Gene Symbol: C15orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207444.2 | NP_997327.1 | 2 | 288 | ||
| GenBank mRNA | AK093014.1 | - | 2 | 288 | ||
| BC119016.1 | - | 2 | 288 | 40108911 | ||
| BC119810.1 | - | 2 | 288 | 40108916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15orf53| Gene Symbol: C15orf53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207444.2 | NP_997327.1 | 2 | 288 | ||
| GenBank mRNA | AK093014.1 | - | 2 | 288 | ||
| BC119016.1 | - | 2 | 288 | 40108911 | ||
| BC119810.1 | - | 2 | 288 | 40108916 | ||