siRNA Details
| Gene Symbol: C18orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272093.2 | NP_001259022.2 | 2 | 500 | ||
| GenBank mRNA | AK126293.1 | - | 2 | 500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C18orf65| Gene Symbol: C18orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272093.2 | NP_001259022.2 | 2 | 500 | ||
| GenBank mRNA | AK126293.1 | - | 2 | 500 | ||