siRNA Details
| Gene Symbol: LOC400661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122073.1 | 1 | 339 | |||
| GenBank mRNA | AK126829.1 | - | 1 | 339 | ||
| BC119774.1 | - | 1 | 279 | 40109466 | ||
| BC119776.1 | - | 1 | 279 | 40109460 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC400661| Gene Symbol: LOC400661 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122073.1 | 1 | 339 | |||
| GenBank mRNA | AK126829.1 | - | 1 | 339 | ||
| BC119774.1 | - | 1 | 279 | 40109466 | ||
| BC119776.1 | - | 1 | 279 | 40109460 | ||