siRNA Details
Gene Symbol: LINC00083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125993.1 | - | 5 | 1831 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00083 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125993.1 | - | 5 | 1831 |