siRNA Details
Gene Symbol: LINC00951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038887.1 | 3 | 2792 | |||
GenBank mRNA | AK123643.1 | - | 3 | 2792 | ||
BC132803.1 | - | 3 | 532 | |||
BC132805.1 | - | 3 | 532 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038887.1 | 3 | 2792 | |||
GenBank mRNA | AK123643.1 | - | 3 | 2792 | ||
BC132803.1 | - | 3 | 532 | |||
BC132805.1 | - | 3 | 532 |