siRNA Details
| Gene Symbol: C7orf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201450.1 | NP_001188379.1 | 2, 3 | 109 | ||
| NM_001201451.1 | NP_001188380.1 | 2, 3 | 109 | |||
| GenBank mRNA | AK124274.1 | - | 2, 3 | 109 | ||
| BC140736.1 | - | 2, 3 | 96 | 9021604 | ||
| BC171740.1 | - | 2, 3 | 96 | 9052960 | ||

C7orf76
Genomic Map