siRNA Details
| Gene Symbol: C9orf170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001709.2 | NP_001001709.1 | 2 | 342 | ||
| GenBank mRNA | AK127445.1 | - | 2 | 342 | ||
| BC132741.1 | - | 2 | 331 | 40146763 | ||
| BC136791.1 | - | 2 | 331 | 9020781 | ||

C9orf170
Genomic Map