siRNA Details
Gene Symbol: SLC22A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004326.4 | NP_001004326.4 | 3 | 647 | ||
NR_033396.1 | 3 | 647 | ||||
XM_011545043.2 | XP_011543345.1 | 3 | 647 | |||
XM_017017774.1 | XP_016873263.1 | 3 | 647 | |||
GenBank mRNA | AK131327.1 | - | 3 | 649 | ||
BC146850.1 | - | 3 | 639 | 9056778 | ||
DY654625.1 | - | 3 | 289 |
