siRNA Details
Gene Symbol: DBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004329.2 | NP_001004329.2 | 4 | 896 | ||
GenBank mRNA | AK131230.1 | - | 6 | 896 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004329.2 | NP_001004329.2 | 4 | 896 | ||
GenBank mRNA | AK131230.1 | - | 6 | 896 |