siRNA Details
| Gene Symbol: ZYG11A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004339.2 | NP_001004339.2 | 5 | 1646 | ||
| NM_001307931.1 | NP_001294860.1 | 4 | 894 | |||
| XM_011541480.2 | XP_011539782.1 | 5 | 1421 | |||
| XM_011541482.2 | XP_011539784.1 | 5 | 1421 | |||
| XM_011541483.2 | XP_011539785.1 | 5 | 1421 | |||
| XM_011541484.1 | XP_011539786.1 | 5 | 1421 | |||
| XM_011541485.1 | XP_011539787.1 | 5 | 1421 | |||
| XM_011541486.1 | XP_011539788.1 | 5 | 1421 | |||
| XM_011541487.2 | XP_011539789.1 | 5 | 1421 | |||
| XM_011541488.1 | XP_011539790.1 | 5 | 1421 | |||
| GenBank mRNA | AY271826.1 | - | 4 | 885 | ||

ZYG11A
Genomic Map