siRNA Details
| Gene Symbol: LOC100506422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004352.2 | NP_001004352.1 | 5 | 628 | ||
| XM_017014175.1 | XP_016869664.1 | 2 | 371 | |||
| GenBank mRNA | AK131322.1 | - | 5 | 629 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506422| Gene Symbol: LOC100506422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004352.2 | NP_001004352.1 | 5 | 628 | ||
| XM_017014175.1 | XP_016869664.1 | 2 | 371 | |||
| GenBank mRNA | AK131322.1 | - | 5 | 629 | ||