siRNA Details
Gene Symbol: STPG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004353.3 | NP_001004353.2 | 4 | 491 | ||
NM_001256699.1 | NP_001243628.1 | 4 | 491 | |||
NM_001256700.1 | NP_001243629.1 | 4 | 491 | |||
NM_001256701.1 | NP_001243630.1 | 4 | 491 | |||
NR_046339.1 | 3 | 356 | ||||
NR_046340.1 | 4 | 411 | ||||
XM_006717113.2 | XP_006717176.1 | 5 | 637 | |||
XM_006717115.2 | XP_006717178.1 | 4 | 494 | |||
GenBank mRNA | AK097565.1 | - | 4 | 474 | ||
BC033939.1 | - | 4 | 471 | 5272537 | ||
BC050094.1 | - | 3 | 312 | |||
BC132726.1 | - | 4 | 491 | 40146748 | ||
BC144308.2 | - | 4 | 491 | 9052828 | ||
BC144311.1 | - | 4 | 491 | 9052831 | ||
BC144314.1 | - | 4 | 491 | 9052834 | ||
DB453255.1 | - | 4 | 457 |
