siRNA Details
| Gene Symbol: LMTK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080434.1 | NP_001073903.1 | 12 | 1290 | ||
| XM_011526411.2 | XP_011524713.1 | 12 | 1621 | |||
| XM_011526412.2 | XP_011524714.1 | 12 | 1588 | |||
| GenBank mRNA | AB067470.1 | - | 12 | 1263 | ||

LMTK3
Genomic Map