siRNA Details
| Gene Symbol: CDKL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009565.1 | NP_001009565.1 | 1 | 152 | ||
| XM_011532815.2 | XP_011531117.1 | 2 | 1568 | |||
| XM_011532816.2 | XP_011531118.1 | 2 | 1568 | |||
| XM_011532817.2 | XP_011531119.1 | 2 | 1568 | |||
| XM_011532819.2 | XP_011531121.1 | 2 | 1568 | |||
| XM_017003981.1 | XP_016859470.1 | 2 | 920 | |||
| XM_017003982.1 | XP_016859471.1 | 2 | 396 | |||
| XM_017003983.1 | XP_016859472.1 | 2 | 1568 | |||
| XM_017003984.1 | XP_016859473.1 | 2 | 1568 | |||
| XM_017003985.1 | XP_016859474.1 | 2 | 1568 | |||
| GenBank mRNA | AY845084.1 | - | 1 | 152 | ||
| AY847283.1 | - | 1 | 152 | |||
| BC144460.1 | - | 1 | 216 | |||

CDKL4
Genomic Map