siRNA Details
| Gene Symbol: SPHK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142601.1 | NP_001136073.1 | 6 | 1352 | ||
| NM_001142602.1 | NP_001136074.1 | 5 | 1397 | |||
| NM_021972.3 | NP_068807.2 | 6 | 1394 | |||
| NM_182965.2 | NP_892010.2 | 6 | 1715 | |||
| XM_005257766.2 | XP_005257823.1 | 6 | 1707 | |||
| GenBank mRNA | AF200328.1 | - | 5 | 978 | ||
| AF238083.1 | - | 6 | 1296 | |||
| AF266756.1 | - | 5 | 1229 | |||
| AK022402.1 | - | 6 | 1352 | |||
| AK292294.1 | - | 6 | 1439 | |||
| AK292614.1 | - | 5 | 1313 | |||
| BC009419.1 | - | 6 | 1382 | 3506924 | ||
| BC030553.1 | - | 6 | 1674 | 5213270 | ||

SPHK1
Genomic Map