siRNA Details
| Gene Symbol: PKDCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138370.2 | NP_612379.2 | 5 | 1375 | ||
| GenBank mRNA | BC007901.2 | - | 3 | 330 | ||
| BC062988.1 | - | 3 | 330 | |||
| BC094697.1 | - | 5 | 783 | 30529646 | ||
| BC110513.1 | - | 5 | 812 | 40031755 | ||
| BC110514.1 | - | 5 | 812 | 40031756 | ||

PKDCC
Genomic Map