siRNA Details
| Gene Symbol: IP6K1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006115.2 | NP_001006115.1 | 5 | 786 | ||
| NM_001242829.1 | NP_001229758.1 | 6 | 1475 | |||
| NM_153273.3 | NP_695005.1 | 6 | 1137 | |||
| GenBank mRNA | AK289772.1 | - | 5 | 942 | ||
| AK295689.1 | - | 5 | 662 | |||
| BC012944.1 | - | 5 | 938 | 3872311 | ||
| D87452.2 | - | 6 | 1124 | |||

IP6K1
Genomic Map