siRNA Details
| Gene Symbol: GRK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004105.2 | NP_001004105.1 | 14 | 1605 | ||
| NM_001004106.2 | NP_001004106.1 | 14 | 1605 | |||
| NM_002082.3 | NP_002073.2 | 14 | 1605 | |||
| XM_006714858.3 | XP_006714921.1 | 12 | 2246 | |||
| XM_006714859.2 | XP_006714922.1 | 14 | 1517 | |||
| XM_011534537.2 | XP_011532839.1 | 13 | 1357 | |||
| XM_011534538.1 | XP_011532840.1 | 7 | 799 | |||
| GenBank mRNA | AF040751.1 | - | 14 | 1448 | ||
| AF040752.1 | - | 14 | 1448 | |||
| AK056697.1 | - | 12 | 1583 | |||
| BC009277.2 | - | 14 | 1545 | 4053197 | ||
| BC017272.1 | - | 14 | 1494 | 4640283 | ||
| L16862.1 | - | 14 | 1507 | |||

GRK6
Genomic Map