siRNA Details
| Gene Symbol: ULK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003565.2 | NP_003556.1 | 27, 28 | 3360 | ||
| XM_011538798.2 | XP_011537100.1 | 27, 28 | 3206 | |||
| XM_011538799.2 | XP_011537101.1 | 27, 28 | 3119 | |||
| GenBank mRNA | AB018265.2 | - | 27, 28 | 3142 | ||
| AF045458.1 | - | 27, 28 | 3360 | |||
| AK095774.1 | - | 1, 2 | 485 | |||

ULK1
Genomic Map