siRNA Details
| Gene Symbol: LIMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204426.1 | NP_001191355.1 | 8, 9 | 1140 | ||
| NM_002314.3 | NP_002305.1 | 9, 10 | 1362 | |||
| GenBank mRNA | AB209557.1 | - | 6, 7 | 834 | ||
| AF134379.1 | - | 9, 10 | 1087 | |||
| AK026544.1 | - | 3, 4 | 398 | |||
| AK290162.1 | - | 10, 11 | 1287 | |||
| AK300382.1 | - | 8, 9 | 1079 | |||
| D26309.1 | - | 10, 11 | 1243 | |||

LIMK1
Genomic Map