siRNA Details
| Gene Symbol: PLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003712.3 | NP_003703.1 | 5 | 738 | ||
| NM_177526.2 | NP_803545.1 | 5 | 697 | |||
| NM_177543.2 | NP_808211.1 | 5 | 805 | |||
| XM_011528396.2 | XP_011526698.1 | 5 | 734 | |||
| GenBank mRNA | AB209696.1 | - | 2 | 5886 | ||
| AF035959.1 | - | 5 | 682 | |||
| AF047760.1 | - | 5 | 682 | |||
| AF056083.1 | - | 5 | 682 | |||
| BC002806.2 | - | 5 | 689 | 3659728 | ||
| BM982485.1 | - | 2 | 583 | |||
| BT007021.1 | - | 5 | 639 | |||

PLPP2
Genomic Map