siRNA Details
| Gene Symbol: DUSP13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007271.1 | NP_001007272.1 | 6 | 1017 | ||
| NM_001007272.1 | NP_001007273.1 | 2 | 292 | |||
| NM_001007273.1 | NP_001007274.1 | 4 | 591 | |||
| NM_001320842.1 | NP_001307771.1 | 5 | 809 | |||
| NM_001320843.1 | NP_001307772.1 | 3 | 387 | |||
| NM_016364.3 | NP_057448.3 | 2 | 150 | |||
| XM_005269883.3 | XP_005269940.1 | 5 | 787 | |||
| XM_005269884.4 | XP_005269941.3 | 2 | 579 | |||
| XM_005269887.1 | XP_005269944.1 | 5 | 889 | |||
| XM_005269890.1 | XP_005269947.1 | 3 | 489 | |||
| XM_011539853.1 | XP_011538155.1 | 6 | 1026 | |||
| XM_011539854.2 | XP_011538156.1 | 4 | 565 | |||
| XM_011539855.1 | XP_011538157.1 | 2 | 268 | |||
| XM_011539856.2 | XP_011538158.1 | 4 | 569 | |||
| XM_017016313.1 | XP_016871802.1 | 2 | 242 | |||
| GenBank mRNA | AB027004.1 | - | 2 | 164 | ||
| AB103375.1 | - | 2 | 164 | |||
| AK057012.1 | - | 3 | 360 | |||
| AK127886.1 | - | 5 | 809 | |||
| AK291891.1 | - | 5 | 889 | |||
| AK291897.1 | - | 4 | 566 | |||
| AK300679.1 | - | 6 | 999 | |||
| AY040091.1 | - | 6 | 981 | |||
| AY674051.1 | - | 6 | 990 | |||
| BC009778.1 | - | 3 | 276 | 4047963 | ||
| BU580940.1 | - | 4 | 558 | |||
| CR457094.1 | - | 1 | 87 | |||

DUSP13
Genomic Map