siRNA Details
| Gene Symbol: DUSP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319658.1 | NP_001306587.1 | 2 | 537 | ||
| NM_001319659.1 | NP_001306588.1 | 3 | 496 | |||
| NM_017823.4 | NP_060293.2 | 3 | 489 | |||
| GenBank mRNA | AB164404.1 | - | 2 | 522 | ||
| AF086553.1 | - | 2 | 337 | |||
| AK000449.1 | - | 3 | 473 | |||
| BC001140.1 | - | 3 | 459 | 3535406 | ||
| BG823348.1 | - | 2 | 519 | |||
| BQ679163.1 | - | 3 | 508 | |||
| DA428735.1 | - | 2 | 540 | |||

DUSP23
Genomic Map