siRNA Details
| Gene Symbol: DGKI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321708.1 | NP_001308637.1 | 2 | 435 | ||
| NM_001321709.1 | NP_001308638.1 | 3 | 213 | |||
| NM_004717.3 | NP_004708.1 | 2 | 435 | |||
| XM_011516682.2 | XP_011514984.2 | 2 | 432 | |||
| XM_017012780.1 | XP_016868269.1 | 2 | 433 | |||
| XM_017012781.1 | XP_016868270.1 | 2 | 432 | |||
| XM_017012782.1 | XP_016868271.1 | 2 | 432 | |||
| XM_017012783.1 | XP_016868272.1 | 2 | 432 | |||
| XM_017012784.1 | XP_016868273.1 | 2 | 432 | |||
| XM_017012788.1 | XP_016868277.1 | 3 | 166 | |||
| GenBank mRNA | DR002813.1 | - | 2 | 440 | ||

DGKI
Genomic Map