siRNA Details
| Gene Symbol: TTBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032538.1 | NP_115927.1 | 7 | 879 | ||
| XM_011514947.2 | XP_011513249.1 | 6 | 669 | |||
| XM_017011364.1 | XP_016866853.1 | 6 | 669 | |||
| XM_017011365.1 | XP_016866854.1 | 6 | 669 | |||
| XM_017011366.1 | XP_016866855.1 | 6 | 669 | |||
| XM_017011367.1 | XP_016866856.1 | 6 | 669 | |||
| GenBank mRNA | AB058758.1 | - | 5 | 448 | ||
| AB218664.1 | - | 8 | 683 | |||
| AK131217.1 | - | 6 | 609 | |||

TTBK1
Genomic Map