siRNA Details
| Gene Symbol: SBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024401.2 | NP_001019572.1 | 2 | 939 | ||
| XM_005255315.3 | XP_005255372.1 | 2 | 973 | |||
| GenBank mRNA | AY874862.2 | - | 2 | 939 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SBK1| Gene Symbol: SBK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024401.2 | NP_001019572.1 | 2 | 939 | ||
| XM_005255315.3 | XP_005255372.1 | 2 | 973 | |||
| GenBank mRNA | AY874862.2 | - | 2 | 939 | ||