siRNA Details
| Gene Symbol: SBK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101401.2 | NP_001094871.2 | 2 | 321 | ||
| XM_006723327.3 | XP_006723390.1 | 3 | 430 | |||
| XM_011527227.2 | XP_011525529.1 | 3 | 429 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SBK2| Gene Symbol: SBK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101401.2 | NP_001094871.2 | 2 | 321 | ||
| XM_006723327.3 | XP_006723390.1 | 3 | 430 | |||
| XM_011527227.2 | XP_011525529.1 | 3 | 429 | |||