siRNA Details
| Gene Symbol: AOC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001158.3 | NP_001149.2 | 1 | 1250 | ||
| NM_009590.2 | NP_033720.2 | 1 | 1250 | |||
| GenBank mRNA | AF081363.1 | - | 1 | 1241 | ||
| BC142641.1 | - | 1 | 1242 | 40147885 | ||
| D88213.1 | - | 1 | 1241 | |||

AOC2
Genomic Map