siRNA Details
| Gene Symbol: AZU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001700.4 | NP_001691.1 | 5 | 728 | ||
| GenBank mRNA | BC044940.1 | - | 4 | 1526 | ||
| BC069495.1 | - | 5 | 723 | 7262247 | ||
| BC093931.1 | - | 5 | 728 | 7939776 | ||
| BC093933.1 | - | 5 | 728 | 7939778 | ||
| BU617927.1 | - | 1 | 205 | |||
| X58794.1 | - | 5 | 701 | |||

AZU1
Genomic Map