siRNA Details
| Gene Symbol: C1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001733.4 | NP_001724.3 | 2 | 221 | ||
| XM_017019915.1 | XP_016875404.1 | 2 | 295 | |||
| GenBank mRNA | AK024951.1 | - | 1 | 921 | ||
| AK222481.1 | - | 2 | 215 | |||
| AK222491.1 | - | 2 | 215 | |||
| AK291313.1 | - | 2 | 243 | |||
| AK298445.1 | - | 2 | 236 | |||
| AK303750.1 | - | 2 | 215 | |||
| BC035220.1 | - | 2 | 221 | 4777940 | ||
| M14058.1 | - | 2 | 225 | |||
| X04701.1 | - | 2 | 213 | |||

C1R
Genomic Map