siRNA Details
| Gene Symbol: CDX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001265.4 | NP_001256.3 | 1 | 860 | ||
| XM_011534875.2 | XP_011533177.1 | 1 | 829 | |||
| XM_011534876.2 | XP_011533178.1 | 1 | 829 | |||
| XM_011534877.2 | XP_011533179.1 | 1 | 829 | |||
| GenBank mRNA | BC014461.1 | - | 1 | 672 | 4867859 | |
| KJ081251.1 | - | 1 | 492 | |||
| U51096.1 | - | 1 | 562 | |||
| Y13709.1 | - | 1 | 858 | |||

CDX2
Genomic Map