siRNA Details
| Gene Symbol: COX5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001862.2 | NP_001853.2 | 2 | 186 | ||
| GenBank mRNA | BC006229.2 | - | 2 | 186 | 3952882 | |
| BT006742.1 | - | 2 | 157 | |||
| CR541727.1 | - | 2 | 157 | |||
| CR541753.1 | - | 2 | 157 | |||
| M19961.1 | - | 2 | 178 | |||

COX5B
Genomic Map