siRNA Details
| Gene Symbol: CRYBA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057093.1 | NP_476434.1 | 4 | 745 | ||
| NM_057094.1 | NP_476435.1 | 5 | 551 | |||
| GenBank mRNA | AF166331.1 | - | 5 | 545 | ||
| BC006285.2 | - | 4 | 745 | 3960073 | ||
| BF726935.1 | - | 5 | 559 | |||
| BT007447.1 | - | 4 | 516 | |||

CRYBA2
Genomic Map