siRNA Details
| Gene Symbol: CST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322.2 | NP_001313.1 | 1 | 101 | ||
| GenBank mRNA | BC062679.1 | - | 1 | 159 | 30350087 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST2| Gene Symbol: CST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322.2 | NP_001313.1 | 1 | 101 | ||
| GenBank mRNA | BC062679.1 | - | 1 | 159 | 30350087 | |