siRNA Details
| Gene Symbol: CST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001900.4 | NP_001891.2 | 2 | 376 | ||
| GenBank mRNA | BC062678.1 | - | 2 | 373 | 30349574 | |
| BC069514.1 | - | 2 | 313 | 7262733 | ||
| CB985957.1 | - | 379 | ||||
| X70377.1 | - | 2 | 313 | |||

CST5
Genomic Map