siRNA Details
| Gene Symbol: CYP2B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000767.4 | NP_000758.1 | 7 | 1055 | ||
| XM_005258569.4 | XP_005258626.1 | 7 | 1151 | |||
| XM_006723050.3 | XP_006723113.1 | 7 | 1151 | |||
| XM_011526546.2 | XP_011524848.1 | 7 | 1154 | |||
| XM_011526547.2 | XP_011524849.1 | 7 | 1152 | |||
| XM_011526548.2 | XP_011524850.1 | 4 | 668 | |||
| XM_011526549.2 | XP_011524851.1 | 5 | 610 | |||
| XM_011526550.2 | XP_011524852.1 | 3 | 463 | |||
| GenBank mRNA | AF182277.1 | - | 7 | 1050 | ||
| AK301620.1 | - | 3 | 452 | |||
| BC067430.1 | - | 7 | 1055 | 7002137 | ||
| BC067431.1 | - | 7 | 1055 | 7002142 | ||
| M29874.1 | - | 7 | 1054 | |||
| X06399.1 | - | 2 | 542 | |||
| X13494.1 | - | 7 | 987 | |||

CYP2B6
Genomic Map