siRNA Details
| Gene Symbol: ADAM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001569.3 | 10 | 1062 | |||
| NR_024106.2 | 10 | 1062 | ||||
| NR_024107.2 | 10 | 1062 | ||||
| NR_073423.1 | 10 | 1062 | ||||
| GenBank mRNA | AK302269.1 | - | 7 | 609 | ||
| AK308250.1 | - | 10 | 926 | |||

ADAM3A
Genomic Map