siRNA Details
| Gene Symbol: SEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052838.4 | NP_443070.5 | 10 | 1025 | ||
| XM_017022991.1 | XP_016878480.1 | 10 | 1005 | |||
| GenBank mRNA | AF085235.1 | - | 9 | 856 | ||
| AL833004.1 | - | 4 | 1791 | |||
| AY034176.1 | - | 9 | 859 | |||
| BC012161.1 | - | 8 | 847 | 4576220 | ||

SEPT1
Genomic Map