siRNA Details
| Gene Symbol: DLST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001933.4 | NP_001924.2 | 8 | 640 | ||
| NR_033814.1 | 7 | 582 | ||||
| NR_045209.1 | 7 | 591 | ||||
| GenBank mRNA | AK289414.1 | - | 8 | 638 | ||
| AK316463.1 | - | 7 | 557 | |||
| AL538322.3 | - | 1 | 80 | |||
| BX248276.1 | - | 7 | 559 | |||
| BX248764.1 | - | 6 | 471 | |||
| BX248774.1 | - | 7 | 526 | |||
| D16373.1 | - | 8 | 589 | |||
| D17297.1 | - | 1 | 692 | |||
| L37418.1 | - | 8 | 577 | |||

DLST
Genomic Map