siRNA Details
| Gene Symbol: DLX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004405.3 | NP_004396.1 | 2 | 686 | ||
| GenBank mRNA | AB208823.1 | - | 1 | 1034 | ||
| AK291367.1 | - | 2 | 836 | |||
| AK297503.1 | - | 2 | 568 | |||
| AK316129.1 | - | 2 | 836 | |||
| BC032558.1 | - | 2 | 692 | 5562689 | ||
| KJ534825.1 | - | 2 | 474 | |||
| L07919.1 | - | 2 | 223 | |||

DLX2
Genomic Map