siRNA Details
| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004943.1 | NP_004934.1 | 3 | 1660 | ||
| GenBank mRNA | BC019266.1 | - | 1 | 743 | ||
| BT006798.1 | - | 1 | 669 | |||
| CD106470.1 | - | 1 | 313 | |||
| L19267.1 | - | 3 | 1251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DMWD| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004943.1 | NP_004934.1 | 3 | 1660 | ||
| GenBank mRNA | BC019266.1 | - | 1 | 743 | ||
| BT006798.1 | - | 1 | 669 | |||
| CD106470.1 | - | 1 | 313 | |||
| L19267.1 | - | 3 | 1251 | |||