siRNA Details
| Gene Symbol: DNAH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206927.1 | NP_001193856.1 | 63, 64 | 9467 | ||
| XM_011514318.2 | XP_011512620.1 | 62, 63 | 9415 | |||
| XM_011514319.2 | XP_011512621.1 | 62, 63 | 9370 | |||
| XM_011514320.2 | XP_011512622.1 | 61, 62 | 9241 | |||
| XM_011514321.1 | XP_011512623.1 | 60, 61 | 8825 | |||
| XM_017010325.1 | XP_016865814.1 | 63, 64 | 9478 | |||
| XM_017010326.1 | XP_016865815.1 | 63, 64 | 9478 | |||
| GenBank mRNA | AF356519.1 | - | 61, 62 | 8961 | ||

DNAH8
Genomic Map