siRNA Details
| Gene Symbol: FOXF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001451.2 | NP_001442.2 | 2 | 1165 | ||
| GenBank mRNA | AK314167.1 | - | 2 | 1172 | ||
| BC089442.1 | - | 2 | 1154 | 30558276 | ||
| U13219.1 | - | 2 | 1140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FOXF1| Gene Symbol: FOXF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001451.2 | NP_001442.2 | 2 | 1165 | ||
| GenBank mRNA | AK314167.1 | - | 2 | 1172 | ||
| BC089442.1 | - | 2 | 1154 | 30558276 | ||
| U13219.1 | - | 2 | 1140 | |||